ENA394767383 Enamine

6-methyl-1H,2H,3H,5H,6H-pyrrolo[2,3-c]pyridin-5-one

  • MDL number MFCD24842864
  • Cas Number : 1443980-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128418
Availability details for
Quantity
  • Availability
  • Add to Cart