ENA394767386 Enamine

2-propyloxane-4-carboxylic acid

  • MDL number MFCD01546614
  • Cas Number : 77554-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128422
Availability details for
Quantity
  • Availability
  • Add to Cart