ENA394767395 Enamine

[1-(aminomethyl)cyclohexyl]methanol

  • MDL number MFCD08063179
  • Cas Number : 2041-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101726

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101726
Availability details for
Quantity
  • Availability
  • Add to Cart