ENA394767406 Enamine

ethyl 4-(2H-1,3-benzodioxol-5-yl)-2,4-dioxobutanoate

  • MDL number MFCD03411533
  • Cas Number : 62849-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128457

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128457
Availability details for
Quantity
  • Availability
  • Add to Cart