ENA394767407 Enamine

2-amino-3-(furan-2-yl)propanoic acid

  • MDL number MFCD06656854
  • Cas Number : 4066-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128458

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128458
Availability details for
Quantity
  • Availability
  • Add to Cart