ENA394767411 Enamine

1-benzofuran-3-carboxamide

  • MDL number MFCD01006723
  • Cas Number : 959304-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128462
Availability details for
Quantity
  • Availability
  • Add to Cart