ENA394767413 Enamine

2-(cyclopropylcarbamoyl)acetic acid

  • MDL number MFCD16692538
  • Cas Number : 1247436-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128464
Availability details for
Quantity
  • Availability
  • Add to Cart