ENA394767415 Enamine

2-(N-hydroxyimino)-N-(3-iodophenyl)acetamide

  • Cas Number : 63608-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128466
Availability details for
Quantity
  • Availability
  • Add to Cart