ENA394767419 Enamine

methyl 2-cyano-2-methylacetate

  • MDL number MFCD00036017
  • Cas Number : 14618-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128472

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P330,P332+P313,P337+P313,P342+P311,P362,P363,P370+P378,P403+P235,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-128472
pictograms GHS07,GHS08
hstatements H227,H302,H312,H315,H319,H332,H334
Availability details for
Quantity
  • Availability
  • Add to Cart