ENA394767420 Enamine

4-methoxy-5-nitro-1H-indazole

  • MDL number MFCD24842873
  • Cas Number : 1443980-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128473
Availability details for
Quantity
  • Availability
  • Add to Cart