ENA394767424 Enamine

3-[(3,4-dimethoxyphenyl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD01185849
  • Cas Number : 468747-44-6
  • Molecular Weight : 251.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128478
Availability details for
Quantity
  • Availability
  • Add to Cart