ENA394767429 Enamine

ethyl 2-(trifluoroacetamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD00411677
  • Cas Number : 171734-43-3
  • Molecular Weight : 321.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128495
Availability details for
Quantity
  • Availability
  • Add to Cart