ENA394767439 Enamine

8-(2,2,2-trifluoroethyl)-1,4-dioxa-8-azaspiro[4.5]decane

  • MDL number MFCD23715977
  • Cas Number : 710350-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128566
Availability details for
Quantity
  • Availability
  • Add to Cart