ENA394767440 Enamine

1-(1-methoxypropan-2-yl)-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD20330786
  • Cas Number : 1344109-81-4
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-methoxy-1-methylethyl)-3-methyl-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C9H14N2O3/c1-6(5-14-3)11-4-8(9(12)13)7(2)10-11/h4,6H,5H2,1-3H3,(H,12,13)
Inchi Key LOCZDXNZJXUXIR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128567
Availability details for
Quantity
  • Availability
  • Add to Cart