ENA394767441 Enamine

2-methoxypyridin-3-ol

  • MDL number MFCD00457990
  • Cas Number : 13472-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128571

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-128571
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart