ENA394767444 Enamine

3-amino-4-ethylbenzene-1-sulfonic acid

  • MDL number MFCD24625440
  • Cas Number : 3694-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268...270
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128574
Availability details for
Quantity
  • Availability
  • Add to Cart