ENA394767448 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-amine

  • MDL number MFCD23826569
  • Cas Number : 1443979-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104357
Availability details for
Quantity
  • Availability
  • Add to Cart