ENA394767449 Enamine

2-(2-methylpropyl)-6-oxopiperidine-4-carboxylic acid

  • MDL number MFCD24842878
  • Cas Number : 1443980-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128579
Availability details for
Quantity
  • Availability
  • Add to Cart