ENA394767453 Enamine

2-propyloxan-4-ol

  • MDL number MFCD19650614
  • Cas Number : 23077-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128585
Availability details for
Quantity
  • Availability
  • Add to Cart