ENA394767455 Enamine

3-sulfanyl-1,2,4-triazin-5-ol

  • MDL number MFCD00047460
  • Cas Number : 626-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104375
Availability details for
Quantity
  • Availability
  • Add to Cart