ENA394767469 Enamine

methyl 6-(aminomethyl)-2-cyclopropylpyridine-3-carboxylate hydrochloride

  • MDL number MFCD24842882
  • Cas Number : 1443980-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128630
Availability details for
Quantity
  • Availability
  • Add to Cart