ENA394767471 Enamine

tert-butyl N-[(3-aminocyclobutyl)methyl]carbamate

  • MDL number MFCD20232875
  • Cas Number : 1334499-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128639
Availability details for
Quantity
  • Availability
  • Add to Cart