ENA394767475 Enamine

3-chloro-2-methylpyridin-4-amine

  • MDL number MFCD09702443
  • Cas Number : 97944-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105097
Availability details for
Quantity
  • Availability
  • Add to Cart