ENA394767482 Enamine

1-(2-methoxyethyl)-3-methyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11643085
  • Cas Number : 1098353-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128664
Availability details for
Quantity
  • Availability
  • Add to Cart