ENA394767483 Enamine

4-chloro-5-nitroquinazoline

  • MDL number MFCD18448299
  • Cas Number : 75224-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128665
Availability details for
Quantity
  • Availability
  • Add to Cart