ENA394767486 Enamine

bicyclo[2.2.2]octane-1-carboxylic acid

  • MDL number MFCD00218755
  • Cas Number : 699-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106638
Availability details for
Quantity
  • Availability
  • Add to Cart