ENA394767490 Enamine

2,1-benzothiazol-5-amine

  • MDL number MFCD18811785
  • Cas Number : 51253-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128672
Availability details for
Quantity
  • Availability
  • Add to Cart