ENA394767494 Enamine

1-(4-fluoro-3-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD17128925
  • Cas Number : 63675-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128676
Availability details for
Quantity
  • Availability
  • Add to Cart