ENA394767498 Enamine

4-methylpiperidine-4-carbonitrile hydrochloride

  • MDL number MFCD12406584
  • Cas Number : 948894-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128680
Availability details for
Quantity
  • Availability
  • Add to Cart