ENA394767506 Enamine

1-{bicyclo[4.2.0]octa-1,3,5-trien-7-yl}ethan-1-amine

  • MDL number MFCD19210096
  • Cas Number : 3264-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128694
Availability details for
Quantity
  • Availability
  • Add to Cart