ENA394767508 Enamine

3-(3-methoxyphenyl)-1,2,4-oxadiazole-5-thiol

  • MDL number MFCD17259823
  • Cas Number : 1339516-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128708
Availability details for
Quantity
  • Availability
  • Add to Cart