ENA394767511 Enamine

6-bromo-8-methyl-1,2-dihydroquinolin-2-one

  • MDL number MFCD11226924
  • Cas Number : 99465-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 278...280
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128714
Availability details for
Quantity
  • Availability
  • Add to Cart