ENA394767514 Enamine

2-acetamido-N-methylbutanediamide

  • MDL number MFCD20709943
  • Cas Number : 36318-48-6
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(acetylamino)-N~1~-methylsuccinamide
Inchi Code 1S/C7H13N3O3/c1-4(11)10-5(3-6(8)12)7(13)9-2/h5H,3H2,1-2H3,(H2,8,12)(H,9,13)(H,10,11)
Inchi Key AOZXXHVRXISMKA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128718
Availability details for
Quantity
  • Availability
  • Add to Cart