ENA394767521 Enamine

5-chloro-1-methyl-3-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD05860084
  • Cas Number : 29938-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107872
Availability details for
Quantity
  • Availability
  • Add to Cart