ENA394767522 Enamine

1,2-bis(benzyloxy)-4-(2-nitroprop-1-en-1-yl)benzene

  • MDL number MFCD02258810
  • Cas Number : 62932-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128732

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128732
Availability details for
Quantity
  • Availability
  • Add to Cart