ENA394767539 Enamine

2-ethyloxane-4-carbaldehyde

  • MDL number MFCD18814949
  • Cas Number : 115602-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128752

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128752
Availability details for
Quantity
  • Availability
  • Add to Cart