ENA394767544 Enamine

4-[3-(piperidin-4-yl)propyl]morpholine

  • MDL number MFCD16693182
  • Cas Number : 933707-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128761

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128761
Availability details for
Quantity
  • Availability
  • Add to Cart