ENA394767545 Enamine

2-(3-methoxyphenyl)-2-(morpholin-4-yl)acetonitrile

  • MDL number MFCD00172615
  • Cas Number : 123567-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128762
Availability details for
Quantity
  • Availability
  • Add to Cart