ENA394767546 Enamine

1,1-dimethyl 2-methylcyclobutane-1,1-dicarboxylate

  • MDL number MFCD21773099
  • Cas Number : 14132-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128767

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128767
Availability details for
Quantity
  • Availability
  • Add to Cart