ENA394767555 Enamine

2-chloro-4,6-dimethyl-3-nitropyridine

  • MDL number MFCD18261551
  • Cas Number : 89793-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128839
Availability details for
Quantity
  • Availability
  • Add to Cart