ENA394767556 Enamine

N-(4-propoxyphenyl)methanesulfonamide

  • MDL number MFCD20189505
  • Cas Number : 1266530-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128851
Availability details for
Quantity
  • Availability
  • Add to Cart