ENA394767557 Enamine

1-(3-cyanophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD17293356
  • Cas Number : 1274470-88-0
  • Molecular Weight : 227.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-cyanophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C12H9N3O2/c1-8-5-11(12(16)17)14-15(8)10-4-2-3-9(6-10)7-13/h2-6H,1H3,(H,16,17)
Inchi Key RZIKVFGCJNRAHX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128852
Availability details for
Quantity
  • Availability
  • Add to Cart