ENA394767570 Enamine

2-(pentylamino)acetamide

  • MDL number MFCD00865293
  • Cas Number : 76990-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128867
Availability details for
Quantity
  • Availability
  • Add to Cart