ENA394767572 Enamine

N,N-dimethyl-4-sulfamoylbenzamide

  • MDL number MFCD11104011
  • Cas Number : 38576-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128880
Availability details for
Quantity
  • Availability
  • Add to Cart