ENA394767578 Enamine

4-(aminomethyl)-1-[2-(cyclopentyloxy)ethyl]pyrrolidin-2-one

  • MDL number MFCD19677542
  • Cas Number : 1339660-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128889
Availability details for
Quantity
  • Availability
  • Add to Cart