ENA394767582 Enamine

1-[2-(diphenylmethoxy)ethyl]piperazine dihydrochloride

  • MDL number MFCD01455758
  • Cas Number : 110407-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114462
Availability details for
Quantity
  • Availability
  • Add to Cart