ENA394767586 Enamine

2-oxononanoic acid

  • MDL number MFCD00090783
  • Cas Number : 13139-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128962
Availability details for
Quantity
  • Availability
  • Add to Cart