ENA394767592 Enamine

1-(furan-2-yl)-3-methoxypropan-1-amine

  • MDL number MFCD16847544
  • Cas Number : 1250282-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128970
Availability details for
Quantity
  • Availability
  • Add to Cart