ENA394767596 Enamine

2-methyl-6-oxopiperidine-4-carboxylic acid

  • MDL number MFCD24842910
  • Cas Number : 36412-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128974

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128974
Availability details for
Quantity
  • Availability
  • Add to Cart