ENA394767600 Enamine

2-(2,3-dichlorophenyl)-2-[(trimethylsilyl)oxy]acetonitrile

  • MDL number MFCD16786508
  • Cas Number : 1250020-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128980

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128980
Availability details for
Quantity
  • Availability
  • Add to Cart